Learn More
Thermo Scientific™ Water, LC-MS Grade
Thermo Scientific™ Water, LC/MS Grade is ultapure water for formulation of solvents for HPLC
68.75 € - 324.00 €
Specifications
Color | Clear, Colorless |
---|---|
Melting Point | 32°F |
Boiling Point | 212°F |
Grade | LC-MS |
Physical Form | Liquid |
Description
Thermo Scientific™ Water, LC/MS Grade is ultapure water for formulation of solvents for HPLC.
An ultrapure, LC/MS-grade preparation with low UV-absorptivity that makes it suitable and reliable for use in HPLC and mass spectrometry applications.
Thermo Scientific Water, LC/MS Grade is specially purified by a proprietary method and treated to ensure lot-to-lot consistency with a low UV absorbance to provide you with most sensitive detection across all wavelengths. Water is 0.2-micron filtered, packaged in solvent-rinsed amber glass bottles and sealed under a nitrogen atmoshere with Teflon TFE-lined fluorocarbon caps for ultimate protection.
Highlights:
- Purity and quality validated with 30 tests for use in LC/MS applications
- 0.2-micron filtered, packed in solvent-rinsed amber glass bottles
- Sealed under nitrogen atmosphere to eliminate absorption of unknown atmospheric gases
Product Details:
General properties of water
- Molecular formula: H2O
- Molecular Weight: 18.01
- CAS number: 7732-18-5
Water, LC/MS Grade is specifically purified and tested to the highest specifications to ensure the integrity of your data, maximize sensitivity in your assay and prolong the life of your equipment.
Specifications
Clear, Colorless | |
212°F | |
Liquid |
32°F | |
LC-MS | |
Pierce™ |
Safety and Handling
Recommended Storage : Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.
For Research Use Only. Not for use in diagnostic procedures.