missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Water, LC-MS Grade

Thermo Scientific™ Water, LC/MS Grade is ultapure water for formulation of solvents for HPLC

Brand:  Thermo Scientific™ 51140

 View more versions of this product

Product Code. 10763295

This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Water, LC/MS Grade is ultapure water for formulation of solvents for HPLC.

An ultrapure, LC/MS-grade preparation with low UV-absorptivity that makes it suitable and reliable for use in HPLC and mass spectrometry applications.

Thermo Scientific Water, LC/MS Grade is specially purified by a proprietary method and treated to ensure lot-to-lot consistency with a low UV absorbance to provide you with most sensitive detection across all wavelengths. Pierce Water is 0.2-micron filtered, packaged in solvent-rinsed amber glass bottles and sealed under a nitrogen atmoshere with Teflon TFE-lined fluorocarbon caps for ultimate protection.

Highlights:

  • Purity and quality validated with 30 tests for use in LC/MS applications
  • 0.2-micron filtered, packed in solvent-rinsed amber glass bottles
  • Sealed under nitrogen atmosphere to eliminate absorption of unknown atmospheric gases

Product Details:

General properties of water

  • Molecular formula: H2
  • Molecular Weight: 18.01
  • CAS number: 7732-18-5

Water, LC/MS Grade is specifically purified and tested to the highest specifications to ensure the integrity of your data, maximize sensitivity in your assay and prolong the life of your equipment.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

6 to 8
32°F
LC-MS
Liquid
Clear, Colorless
212°F
1 L
Pierce™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Water, LC-MS Grade > Water; 1L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.