Learn More
Gilson™ PIPETMAN L 4-Pipette Kit

The latest manual pipette from Gilson™, PIPETMAN L combines the light and responsive piston spring of the PIPETMAN G with a new body shape designed to improve the ergonomics of the pipette.
Brand: Gilson™ F167370
1028.15 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
- Locking system design which guarantees the elimination of user interaction volume drift during pipetting
- A truly user-friendly pipette for modern busy laboratories
- Contains 4 pipettes (2L, 20L, 200L, 1000L), conveniently packaged with 3 racks of PIPETMAN DIAMOND Tips (10L, 200L, 1000L), 4 single pipette holders, 1 Gilson Guide to Pipetting
- 2D identification code for easy traceability

Specifications
Diamond | |
Pipetman L | |
0.2-2 μL, 100-1000 μL, 2-20 μL, 20-200 μL |
1 | |
Adjustable Volume Manual Pipette | |
3 years |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.