Learn More
Eppendorf™ Pipette Carousel 2
Carries up to six manual single or multi-channel Eppendorf pipettes.
Brand: Eppendorf™ 3116000015
168.99 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
Carries up to six manual single or multi-channel Eppendorf pipettes.
- Includes rotatable stand with completely new design that holds all current manual pipettes
- Pipette holders are interchangeable, no tools required
- Robust design that securely holds and protects pipettes
- Handle for easy transport from bench to bench
- When used with optional pipette holder, the carousel can carry the Multipette
Specifications
Manual Single or Multi-channel Eppendorf Pipettes (6) | |
6 |
Product Content Correction
Product Title
Eppendorf™ Pipette Carousel 2 >
Spot an opportunity for improvement?Share a Content Correction