Learn More
Steris™ Plain Top Closure Pouches
For use in porous load pre-vacuum autoclaves as well as gamma sterilizers. Steris Corporation™ Plain Top Closure Pouches include a double folded bottom seal for extra integrity.
Brand:  Steris™ MPC
 View more versions of this product
Product Code. 15661964
  • 174.00 € / Pack of 250
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Manufactured from high quality wet strength MG bleached kraft paper
  • Double folded bottom seal for extra integrity
  • Opening along width
  • Conforms to BS 6257
Specifications
Specifications
Autoclavable
Plain
250/Pk.
300 mm
530 mm
BS 6257
Medical Grade Kraft Paper (Bleached)
300 x 75 x 530mm
75 mm