Learn More
Steris™ Plain Top Closure Pouches
For use in porous load pre-vacuum autoclaves as well as gamma sterilizers. Steris Corporation™ Plain Top Closure Pouches include a double folded bottom seal for extra integrity.
174.00 € - 224.00 €
Specifications
Autoclavable Autoclavable
Certifications/Compliance BS 6257
Closure Type Plain
Material Medical Grade Kraft Paper (Bleached)
Quantity 250/Pk.
View More Specs
Products 2
Product Code Brand Dimensions (L x W x H) Price Quantity & Availability  
Product Code Brand Dimensions (L x W x H) Price Quantity & Availability  
15661964
View Documents
Steris™
MPC
300 x 75 x 530mm
174.00 €
Pack of 250
In Stock
to see stock.
Edge
Add to basket
15681964
View Documents
Steris™
T35PC
380 x 125 x 610mm
224.00 €
Pack of 250
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • Manufactured from high quality wet strength MG bleached kraft paper
  • Double folded bottom seal for extra integrity
  • Opening along width
  • Conforms to BS 6257
Specifications
Specifications
Autoclavable
Plain
250/Pk.
BS 6257
Medical Grade Kraft Paper (Bleached)