missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Plastic Analog/Dial Caliper

These easy-to-read dial calipers offer accurate measurements at an economical price.

Brand:  Cole-Parmer™ 97100-10

Product Code. 11759539

  • 112.00 € / Each
Estimated Shipment: 17-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Made of fiberglass-reinforced plastic with beveled jaws.
  • Inside, outside, depth, and step measurements
  • Bar has inch and mm graduations on the front and a fractional conversion table on the back
  • No-glare lens for easy reading.
Specifications

Specifications

Analog/Dial Caliper, Plastic; 0 to 150 mm
0 to 150 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole-Parmer™ Plastic Analog/Dial Caliper > Range: 0 to 150mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.