missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Plastic Coated Steel Lockout Device Cable

Lock out gate valves, gas cylinders, electrical devices and more with this Plastic Coated Steel Lockout Device Cable.

Brand:  Brady™ 800114

Product Code. 11776685

  • 69.85 € / Each
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Provide extra information by combining the device with a tag (not included).
  • Dimensions: 16.6 x 148.5 x 50.2 mm (D x W x H)
Specifications

Specifications

Lockout Cable
Red
1 Ea.
CABLO-10
Cable Lockout Device and 3 m cable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brady™ Plastic Coated Steel Lockout Device Cable > Plastic Coated Steel Lockout Device Cable; Material: Glass-Filled Polypropylene and Plastic Coated Steel

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.