Learn More
Multiroir Controlec™ Plastic Mobile Storage Cart
The Multiroir multi-drawer storage cart is ideal for general storage of items that can easily be seen through the translucent plastic drawers. The cart is on casters so it can easily be moved around the workspace.
Brand:  Multiroir Controlec™ 18105
Product Code. 16437570
  • 69.40 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Black housing on casters
  • Translucent drawers make objects easily visible
  • Lightweight
  • Resistant to shocks, cleaning products and temperature variations
  • Made in France
Specifications
Specifications
Not Autoclavable
Black
40 x 29 x 67 cm (with casters)
Polypropylene
3
15L drawers
Black tower with 3 transparent drawers
General storage
4 casters, 360°
2.4 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Multiroir Controlec™ Plastic Mobile Storage Cart > Capacity (Metric): 3 x drawers