Learn More
Thermo Scientific™ Water, nuclease-free
Optimize molecular biology applications with this deionized and 0.22μm membrane-filtered nuclease-free water.
Brand: Thermo Scientific™ R0582
37.81 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Nuclease-free water is deionized and 0.22 μm membrane-filtered nuclease-free water. It is ideal for all .
Ideal for all molecular biology applications.- Free of endo-, exodeoxyribonucleases, ribonucleases and phosphatases

Specifications
7.6 | |
Bottle | |
30 mL |
Molecular Biology | |
Not DEPC-Treated |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.