Learn More
Thermo Scientific™ Plates and Modules with Covalent Binding Surfaces
Reduce the amount of reagents used in the lab by using these plates and modules with covalent binding surfaces.
Brand:  Thermo Scientific™ 436012
 View more versions of this product
Product Code. 10029152
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Reduce the amount of reagents used in the lab with Thermo Scientific™ Plates and Modules with Covalent Binding Surfaces.
Covalent Surfaces (CovaLink™ and Immobilizer Amino)
  • Minimal leaching
  • Withstand vigorous washing
  • Coating with lower amounts of reagent may be possible
  • Control of orientation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
Fluorescence
30 Cases, 5 Pk.
Black
Polystyrene
384
Non-sterile
120 μL
30
Unspecified
Peptides
Without Lid
16 x 24
Covalent
Solid plate 384-Well
120 μL
50 μL
Amine
Flat Bottom (F-well)
5
30/Cs.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.