Learn More
Thermo Scientific™ Plates and Modules with Covalent Binding Surfaces

Reduce the amount of reagents used in the lab by using these plates and modules with covalent binding surfaces.
487.83 € - 1094.04 €
Specifications
Material | Polystyrene |
---|---|
No. per Case | 30 |
No. per Pack | 5 |
Plate Blocking | Unspecified |
Target Molecule | Peptides |
Product Code | Brand | Color | Volume (Metric) Working | Product Type | Surface Treatment | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Volume (Metric) Working | Product Type | Surface Treatment | Price | Quantity & Availability | ||||||
10316872
![]() |
Thermo Scientific™
436006 |
Clear | 100 μL | Microplate | Covalent Binding |
672.66 €
Pack of 30 |
In Stock
Log in to see stock. |
||||||
10432612
|
Thermo Scientific™
436007 |
White | 100 μL | Solid plate 96-Well | Covalent Binding |
969.60 €
Pack of 30 |
In Stock
Log in to see stock. |
||||||
10052232
![]() |
Thermo Scientific™
436008 |
Black | 100 μL | Microplate | Amine |
957.48 €
Pack of 30 |
In Stock
Log in to see stock. |
||||||
10062242
|
Thermo Scientific™
436009 |
Clear | 50 μL | Solid plate 384-Well | Amine | N/A | N/A | N/A | |||||
10029152
|
Thermo Scientific™
436012 |
Black | 50 μL | Solid plate 384-Well | Amine | N/A | N/A | N/A | |||||
10432802
![]() |
Thermo Scientific™
436013 |
Clear | 100 μL | Microplate | Amine |
770.63 €
Pack of 30 |
In Stock
Log in to see stock. |
||||||
10646083
![]() |
Thermo Scientific™
436023 |
Clear | 100 μL | Microplate | Amine |
770.63 €
Pack of 30 |
In Stock
Log in to see stock. |
||||||
10499762
|
Thermo Scientific™
478042 |
Clear | 100 μL | Solid Module F8 (x12) in Frame | Covalent Binding |
487.83 €
Pack of 30 |
In Stock
Log in to see stock. |
||||||
Description
Reduce the amount of reagents used in the lab with Thermo Scientific™ Plates and Modules with Covalent Binding Surfaces.
Covalent Surfaces (Thermo Scientific™ CovaLink™ and Immobilizer Amino)
- Minimal leaching
- Withstands vigorous washing
- Coating with lower amounts of reagent may be possible
- Control of orientation
Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.
Specifications
Polystyrene | |
5 | |
Peptides |
30 | |
Unspecified |
Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.