Documents
Scientific Resources
Application Notes
- 1
- 2
Reduce the amount of reagents used in the lab by using these plates and modules with covalent binding surfaces.
Brand: Thermo Scientific™ 436013
593.39 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.
763.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Reduce the amount of reagents used in the lab with Thermo Scientific™ Plates and Modules with Covalent Binding Surfaces.
Covalent Surfaces (Thermo Scientific™ CovaLink™ and Immobilizer Amino)
Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.
Colorimetric | |
Covalent | |
Microplate | |
100 μL | |
Amine | |
30 | |
Unspecified |
30 Cases | |
Clear | |
Polystyrene | |
Non-sterile | |
Round, Flat Bottom (F-well) | |
5 | |
Peptides |
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
185001 | Certificate of Compliance | 22/02/2025 | 436013 |
185000 | Certificate of Compliance | 08/02/2025 | 436013 |
169888 | Certificate of Compliance | 06/12/2024 | 436013 |
167454 | Certificate of Compliance | 03/12/2024 | 436013 |
181752 | Certificate of Compliance | 03/12/2024 | 436013 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.