missing translation for 'onlineSavingsMsg'
Learn More

Platinum, 5% on alumina powder, reduced, Thermo Scientific Chemicals

116.58 € - 1740.66 €

Chemical Identifiers

CAS 7440-6-6
MDL Number MFCD03458043
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15461977
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044222.06
5 g
116.58 €
5g
Please to purchase this item. Need a web account? Register with us today!
11379728
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044222.14
25 g
480.39 €
25g
Please to purchase this item. Need a web account? Register with us today!
11389728
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044222.22
100 g
1740.66 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Platinum on Alumina (aluminum oxide) is catalyst material with applications in synthetic chemistry such as hydrogenation of functional groups. catalyst is recommended for selective hydrogenation reactions. The particle size of the catalyst is ideal for allowing fast separation from the reaction mixture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-6-6
MFCD03458043

Specifications

7440-6-6
MFCD03458043
Powder
Powder
Insoluble in water,moderately soluble in strong mineral acids.
Platinum, 5% on alumina powder
Videos
Safety and Handling

Safety and Handling

H228-H315-H319-H335 : Flammable solid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only