missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ Taq DNA Polymerase, DNA-free

Invitrogen Platinum Taq DNA Polymerase, DNA-free, is manufactured using a novel, closed single-use system together with stringent quality-control testing to minimize the risk of contamination with bacterial and human DNA.

Brand:  Invitrogen™ 15966005

261.00 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 16315062

  • 348.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Invitrogen Platinum Taq DNA Polymerase, DNA-free, is manufactured using a novel, closed single-use system together with stringent quality-control testing to minimize the risk of contamination with bacterial and human DNA. This enzyme is an ideal choice for PCR-based applications requiring the high sensitivity without false-positive results from reagent-borne contamination. This DNA-free enzyme provides the same high level of performance and lot-to-lot consistency as standard Platinum Taq DNA Polymerase.

Platinum Taq DNA Polymerase, DNA-free, is a recombinant Taq polymerase complexed with a proprietary antibody that blocks polymerase activity at ambient temperatures and dissociates after the initial denaturation step at 94°C. This automatic hot start provides increased sensitivity, specificity, and yield, while allowing convenient reaction assembly at room temperature. Just as with Taq DNA Polymerase, Platinum Taq DNA Polymerase, DNA-free, has a non-template–dependent terminal transferase activity that adds a 3' deoxyadenosine to product ends and has a 5' to 3' exonuclease activity.

Features

  • Certified low DNA contamination level (≤0.01 copy of bacterial gDNA per enzyme unit)
  • Confidence in qualitative and quantitative detection of low-abundance microorganisms due to signal absence with no-template/reagents-only PCR controls
  • Same performance as Platinum Taq DNA Polymerase manufactured using conventional process

Applications

  • Amplification of DNA from complex genomic and viral templates
  • Hot start PCR
  • RT-PCR
TRUSTED_SUSTAINABILITY
Specifications

Specifications

• Platinum Taq DNA Polymerase, DNA-free, 100 μL
• 10X PCR Buffer (without magnesium), DNA-free, 1.25 mL
• 50 mM MgCl2, DNA-free (1 mL)

Store at -20°C in a non-frost-free freezer.
Low
Standard
500 Units
Hot-start PCR
1X
400 Reactions
Separate Components
DNA
Tube
Platinum Taq DNA Polymerase
Taq DNA Polymerase
Dry Ice
5 U/μL
Built-In Hot Start
3'-A
5 kb or less
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2794717 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number15966005
Lot Number2794696 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number15966005
Lot Number2796231 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number15966005
Lot Number2826357 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number15966005
Lot Number2826421 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number15966005
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Platinum™ Taq DNA Polymerase, DNA-free > 400 Reactions

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.