Learn More
Invitrogen™ Platinum™ Taq DNA Polymerase, DNA-free
Invitrogen Platinum Taq DNA Polymerase, DNA-free, is manufactured using a novel, closed single-use system together with stringent quality-control testing to minimize the risk of contamination with bacterial and human DNA.
348.00 € - 1526.00 €
Specifications
Format | Tube |
---|---|
GC-Rich PCR Performance | Low |
Polymerase | Platinum Taq DNA Polymerase |
Reaction Speed | Standard |
Product Type | Taq DNA Polymerase |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
16315062
|
Invitrogen™
15966005 |
400 Reactions |
348.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16325062
|
Invitrogen™
15966025 |
2000 Reactions |
1526.00 €
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Invitrogen Platinum Taq DNA Polymerase, DNA-free, is manufactured using a novel, closed single-use system together with stringent quality-control testing to minimize the risk of contamination with bacterial and human DNA. This enzyme is an ideal choice for PCR-based applications requiring the high sensitivity without false-positive results from reagent-borne contamination. This DNA-free enzyme provides the same high level of performance and lot-to-lot consistency as standard Platinum Taq DNA Polymerase.
Platinum Taq DNA Polymerase, DNA-free, is a recombinant Taq polymerase complexed with a proprietary antibody that blocks polymerase activity at ambient temperatures and dissociates after the initial denaturation step at 94°C. This automatic hot start provides increased sensitivity, specificity, and yield, while allowing convenient reaction assembly at room temperature. Just as with Taq DNA Polymerase, Platinum Taq DNA Polymerase, DNA-free, has a non-template–dependent terminal transferase activity that adds a 3' deoxyadenosine to product ends and has a 5' to 3' exonuclease activity.
Features
- Certified low DNA contamination level (≤0.01 copy of bacterial gDNA per enzyme unit)
- Confidence in qualitative and quantitative detection of low-abundance microorganisms due to signal absence with no-template/reagents-only PCR controls
- Same performance as Platinum Taq DNA Polymerase manufactured using conventional process
Applications
- Amplification of DNA from complex genomic and viral templates
- Hot start PCR
- RT-PCR
Specifications
Tube | |
Platinum Taq DNA Polymerase | |
Taq DNA Polymerase | |
Hot-start PCR | |
1X | |
3'-A | |
5 kb or less |
Low | |
Standard | |
Dry Ice | |
5 U/μL | |
Built-In Hot Start | |
Separate Components | |
DNA |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.