missing translation for 'onlineSavingsMsg'
Learn More

VEE GEE Plato Hydrometer with Thermometer

The Plato Hydrometer with Thermometer is calibrated to show the concentration of extract as a percentage by weight in brewery wort.

Brand:  VEE GEE

 View more versions of this product

Product Code. 17846483

This item is not returnable. View return policy

Description

Description

  • Calibration for use at 20°C/68°F
  • Thermometer filled with SafetyBLUE liquid
Specifications

Specifications

Plato Hydrometer
36 cm
Glass
0°C to 50°C
0.1°
7.5 to 16° Plato
32°F to 122°F
20°C (68°F)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.