missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Pluronic™ F-127, low UV absorbance

Pluronic™ F-127, low UV absorbance

Brand:  Molecular Probes™ P6867

48.64 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.


49.21 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 11529316

  • 64.75 € / 2g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Pluronic F-127 is a nonionic, surfactant polyol (molecular weight approximately 12,500 daltons) that has been found to facilitate the solubilization of water-insoluble dyes and other materials in physiological media.1 Pluronic F-127 is commonly used to help disperse the acetoxymethyl (AM) esters of our ion indicators as well as our cell tracer dyes such as CFDA-SE.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Powder
2 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2984203 Certificate TypeCertificate of Analysis Date16/08/2024 Catalog NumberP6867
Lot Number2712262 Certificate TypeCertificate of Analysis Date31/08/2023 Catalog NumberP6867
Lot Number2519456 Certificate TypeCertificate of Analysis Date26/09/2022 Catalog NumberP6867
Lot Number2421167 Certificate TypeCertificate of Analysis Date21/12/2021 Catalog NumberP6867
Lot Number2309142 Certificate TypeCertificate of Analysis Date20/07/2021 Catalog NumberP6867
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molecular Probes™ Pluronic™ F-127, low UV absorbance > 2 g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.