missing translation for 'onlineSavingsMsg'
Learn More

Cytiva PlusOne™ Reagents

High quality electrophoresis reagents. Cytiva Life Sciences™ PlusOne Reagents are a low toxicity, environmentally safe solvent for high quality electrophoresis.

Brand:  Cytiva 17-1332-01


 View more versions of this product

Product Code. 10380315

  • 360.00 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Urea

  • Provides highest resolution and the cleanest results for IEF performed under denaturing conditions
  • Urea solubilizes and unfolds most proteins to their fully random conformation, with all ionizable groups exposed to solution
  • 8M urea included in the 2-D sample solution
Repel-Silane ES
Low toxicity and environmentally safe solvent
  • Inhibits sticking of polyacrylamide gels, agarose gels, and nucleic acids to glass surfaces
  • Bind-Silane
    • Used for covalently attach polyacrylamide gels to a glass surface
    • Firmly attaches to the glass during staining and drying procedures

    Urea:

    • Molecular Weight: 60.06
    • Physical state: Solid
    • Color: White
    • Pack of 500g
    Repel-Silane ES
    • Molecular Weight: 129.06
    • Physical state: Liquid
    • Color: Colorless
    • Pack of 500mL
    Bind-Silane
    • Molecular Weight: 248.4
    • Physical state: Liquid
    • Color: Pale
    • Pack of 25mL

    IEF, PAGE, sequencing

    Certifications

    Suitable for DIGE

    TRUSTED_SUSTAINABILITY
    Specifications

    Specifications

    2530-85-0
    Colorless
    Urea
    Used in IEF, PAGE and Sequencing
    500 mL
    PlusOne Repel-Silane ES
    2%
    Liquid
    129.06
    Dry and dark at room temperature
    Product Suggestions

    Product Suggestions

    Videos
    Safety and Handling

    Safety and Handling

    Signal Word
    • Warning

    Hazard Category
    • Acute toxicity Category 4
    • Flammable liquid Category 3
    • Acute toxicity Category 4
    • LONG-TERM AQUATIC HAZARD Chronic 4
    • Flammable liquid Category 3
    • Reproductive toxicity Category 2
    • LONG-TERM AQUATIC HAZARD Chronic 4
    • Reproductive toxicity Category 2

    Hazard Statement
    • H226-Flammable liquid and vapour.
    • H302 + H312-Harmful if swallowed or in contact with skin.
    • H302-Harmful if swallowed.
    • H361f-Suspected of damaging fertility.
    • H312-Harmful in contact with skin.
    • H413-May cause long lasting harmful effects to aquatic life.

    Precautionary Statement
    • P201-Obtain special instructions before use.
    • P280-Wear protective gloves/protective clothing/eye protection/face protection.
    • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
    • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
    • P273-Avoid release to the environment.
    • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
    • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
    • P235-Keep cool.
    • P501-Dispose of contents/container to .

    Supplemental information
    • MIXTURE LIST-Contains : octamethylcyclotetrasiloxane, dimethyldichlorosilane

    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.