missing translation for 'onlineSavingsMsg'
Learn More

Cytiva PlusOne™ Reagents

High quality electrophoresis reagents. Cytiva Life Sciences™ PlusOne Reagents are a low toxicity, environmentally safe solvent for high quality electrophoresis.

63.93 € - 360.00 €

Specifications

CAS 2530-85-0
View More Specs

Products 3
Product Code Brand Quantity Form Price Quantity & Availability  
Product Code Brand Quantity Form Price Quantity & Availability  
11515005
View Documents
Cytiva
17-1319-01
500 g Solid
106.19 €
500g
In Stock
to see stock.
Add to basket
 
11585005
View Documents
Cytiva
17-1330-01
25 mL Liquid This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10380315
View Documents
Cytiva
17-1332-01
500 mL Liquid
360.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Urea

  • Provides highest resolution and the cleanest results for IEF performed under denaturing conditions
  • Urea solubilizes and unfolds most proteins to their fully random conformation, with all ionizable groups exposed to solution
  • 8M urea included in the 2-D sample solution
Repel-Silane ES
Low toxicity and environmentally safe solvent
  • Inhibits sticking of polyacrylamide gels, agarose gels, and nucleic acids to glass surfaces
  • Bind-Silane
    • Used for covalently attach polyacrylamide gels to a glass surface
    • Firmly attaches to the glass during staining and drying procedures

    Urea:

    • Molecular Weight: 60.06
    • Physical state: Solid
    • Color: White
    • Pack of 500g
    Repel-Silane ES
    • Molecular Weight: 129.06
    • Physical state: Liquid
    • Color: Colorless
    • Pack of 500mL
    Bind-Silane
    • Molecular Weight: 248.4
    • Physical state: Liquid
    • Color: Pale
    • Pack of 25mL

    IEF, PAGE, sequencing

    Certifications

    Suitable for DIGE

    Specifications

    Specifications

    2530-85-0
    Videos
    Safety and Handling

    Safety and Handling

    missing translation for 'shelfLife' : 12 Months

    SDS
    Documents

    Documents

    Product Certifications
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.