missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva PlusOne™ Reagents
High quality electrophoresis reagents. Cytiva Life Sciences™ PlusOne Reagents are a low toxicity, environmentally safe solvent for high quality electrophoresis.
Brand: Cytiva 17-1319-01
This item is not returnable.
View return policy
Description
Urea
- Provides highest resolution and the cleanest results for IEF performed under denaturing conditions
- Urea solubilizes and unfolds most proteins to their fully random conformation, with all ionizable groups exposed to solution
- 8M urea included in the 2-D sample solution
Low toxicity and environmentally safe solvent
- Used for covalently attach polyacrylamide gels to a glass surface
- Firmly attaches to the glass during staining and drying procedures
Urea:
- Molecular Weight: 60.06
- Physical state: Solid
- Color: White
- Pack of 500g
- Molecular Weight: 129.06
- Physical state: Liquid
- Color: Colorless
- Pack of 500mL
- Molecular Weight: 248.4
- Physical state: Liquid
- Color: Pale
- Pack of 25mL
IEF, PAGE, sequencing
Certifications
Suitable for DIGE
Specifications
> 132°C | |
White | |
Solid | |
132°C | |
500 g |
2530-85-0 | |
Urea | |
IEF, PAGE, sequencing | |
60.06 | |
Dry at 5 - 30ᵒC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction