Learn More
Plum™ Plutect Dual Skin Cream
Skin care cream for use before and during work with alternating work processes involving water-based and other substances. May also be applied as replenishing after-work skin cream.
Brand: Plum™ P2503
This item is not returnable.
View return policy
Description
Specifications
Skin Care Cream | |
Bag-in-box | |
0.7 L |
MP2000 dispenser | |
8 Pk. |
Product Content Correction
Product Title
Plum™ Plutect Dual Skin Cream > Packaging: 8 x 0.7 litre bag-in-box
Spot an opportunity for improvement?Share a Content Correction