Learn More
Plum™ Plutect Dual Skin Cream
Skin care cream for use before and during work with alternating work processes involving water-based and other substances. May also be applied as replenishing after-work skin cream.
102.00 € - 222.00 €
Specifications
Product Type Skin Care Cream
View More Specs
Products 2
Product Code Brand Packaging Price Quantity & Availability  
Product Code Brand Packaging Price Quantity & Availability  
15731137
View Documents
Plum™
P2541
Tube
102.00 €
Pack of 18
Estimated Shipment: 11-08-2025
to see stock.
Add to basket
15721137
View Documents
Plum™
P2503
Bag-in-box
222.00 €
Pack of 8
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
Description
Description
  • Effective protection relative to water-based and other substances
  • Perfume-, colour- and paraben-free
  • Resistant to 20 out of 20 substances in Suskind test/winner of BUS test 2007
  • Quickly absorbed, eases skin cleaning
  • Vaseline and zinc oxide provides skin protection and reduces skin irritation
  • Specifications
    Specifications
    Skin Care Cream
    Videos
    SDS
    Documents
    Documents
    Product Certifications
    Product Content Correction
    Product Title
    Plum™ Plutect Dual Skin Cream