missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ PMP Class A Measuring Flask
PMP flask with polypropylene stopper
22.11 € - 50.45 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
17470144
|
BRAND™
36178 |
250 mL |
49.10 €
Pack of 2 |
In Stock
Log in to see stock. |
|||||
10465243
|
BRAND™
36178 |
250 mL | N/A | ||||||
17480144
|
BRAND™
36180 |
500 mL |
50.45 €
Pack of 2 |
In Stock
Log in to see stock. |
|||||
10017602
|
BRAND™
36180 |
500 mL |
22.80 €
Each |
In Stock
Log in to see stock. |
|||||
10766833
|
BRAND™
36182 |
1000 mL |
29.75 €
Each |
In Stock
Log in to see stock. |
|||||
Description
Specifications
PMP | |
PP stopper | |
Autoclavable | |
A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title