missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PolarScreen™ ER Alpha Competitor Assay, Green

1808.00 € - 2405.00 €

Specifications

Detection Method Fluorescence
For Use With (Equipment) Fluorescent Imager
Format 384-well Plate
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15697848
View Documents
Thermo Scientific™
A15883
800 Assays
1808.00 €
Pack of 800
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
 
15340186
View Documents
Thermo Scientific™
A15882
2500 Assays
2405.00 €
Each
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
Description

Description

  • The PolarScreen™ ER (Estrogen Receptor) Alpha Competitor Assay Kit, Green, provides a sensitive and efficient method for high-throughput, fluorescence polarization-based screening of potential ER alpha ligands. The kit uses insect cell-expressed, full-length, native (untagged) human estrogen receptor alpha (available separately, Cat. No. A15674) and a novel, high-affinity, fluorescent estrogen ligand (Fluormone ES2 Green) in a homogenous, mix-and-read assay format.
  • This kit produces a green color read-out; a red kit is also available. Both kits suitably determine the IC50s of competitive compounds. Color choice depends on the spectral properties of the compounds to be screened and the available instrumentation.
  • Complete & ready-to-use out of the box - all reagents included, just add your test compounds
  • High throughput compatible - 800 x 20 μL fluorescence polarization assays in a 384-well plate
  • Convenient - available in bulk sizes for larger screening needs
  • Includes:
    • 2 tubes full-length ER Alpha Recombinant Protein (store at -68 to -85°C)
    • 1 tube of Fluormone ES2 Green Tracer (store at -5 to -30°C)
    • 1 bottle of ES2 Screening Buffer (store at -5 to -30°C)
Specifications

Specifications

Fluorescence
Fluorescent Imager
384-well Plate
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.