Learn More
MP Biomedicals™ Poly-L-Lysine Hydrochloride
Poly-L-Lysine Hydrochloride is a cellular attachment factor in culture applications.
Brand:  MP Biomedicals™ 0215269080
 View more versions of this product
Product Code. 17338583
  • 463.00 € / 100mg
Explore more special offers
Description
Description
  • Poly-L-lysine is a nonspecific attachment factor for cells useful in promoting cell adhesion to solid substrates.
  • Poly-L-lysine enhances electrostatic interaction between negatively charged ions of the cell membrane and the culture surface.
  • When adsorbed to the culture surface, poly-L-lysine increases the number of positively charged sites available for cell binding.
  • Polymers of both D- and L-lysine are used to coat solid surfaces.
  • Poly-L-lysine has been reported to improve the protein coating of ELISA plates.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
26124-78-7
-20°C
Cellular attachment factor in culture applications
100 mg
Whitish
Powder
>20,000 g/mol
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
MP Biomedicals™ Poly-L-Lysine Hydrochloride > Quantity: 100 mg
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.