missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 145,000 to 180,000, Thermo Scientific Chemicals
21.51 € - 222.00 €
Chemical Identifiers
CAS | 9002-89-5 |
---|---|
Molecular Formula | (C2H4O)n |
Molecular Weight (g/mol) | 44.05 |
MDL Number | MFCD00081922 |
InChI Key | IMROMDMJAWUWLK-UHFFFAOYSA-N |
Synonym | vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval |
PubChem CID | 11199 |
IUPAC Name | ethenol |
SMILES | OC(-*)C-* |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15254804
|
Thermo Scientific Acros
450150250 |
25 g | Plastic bottle |
21.51 €
25g |
In Stock
Log in to see stock. |
||||
15234804
|
Thermo Scientific Acros
450155000 |
500 g | Plastic bottle |
131.00 €
500g |
Estimated Shipment: 12-12-2024
Log in to see stock. |
||||
15224804
|
Thermo Scientific Acros
450150010 |
1 kg | Plastic bottle |
222.00 €
1kg |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
9002-89-5 | |
44.05 | |
IMROMDMJAWUWLK-UHFFFAOYSA-N | |
11199 | |
OC(-*)C-* |
(C2H4O)n | |
MFCD00081922 | |
vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval | |
ethenol |
Specifications
9002-89-5 | |
(C2H4O)n | |
vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval | |
IMROMDMJAWUWLK-UHFFFAOYSA-N | |
ethenol | |
11199 |
230.0°C to 240.0°C | |
MFCD00081922 | |
Solubility in water: soluble in hot water. | |
OC(-*)C-* | |
44.05 | |
Poly(vinyl alcohol) |
Safety and Handling
Product Identifier
- Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 145000-180000
RUO – Research Use Only