Learn More
Masterflex™ Polycarbonate Flowmeters with Valve
Brand:  Masterflex™ 32900-44
 View more versions of this product
Product Code. 15233767
  • 131.00 € / Each
Estimated Shipment: 04-04-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Variable Area Flowmeter
Stainless Steel
0.1 to 1.2 LPM
±1%
±4% (Full Scale)
100 psig
Precision-adjusting valve for accurate, leak-free flow control
65°C