missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Polycarbonate Luer Fittings

Designed to work with microtubing and luer-type syringes. Standard-sized connecting ends make these luer fittings completely interchangeable for surefire installations.

Brand:  Masterflex™ 45501-60

 View more versions of this product

Product Code. 11746008

  • 35.20 € / Pack of 25
Estimated Shipment: 07-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Designed to work with microtubing and luer-type syringes. Standard-sized connecting ends make these luer fittings completely interchangeable for surefire installations.

  • Manufactured from 100% virgin raw materials
  • Free of contaminants and ideal for syringes or small tubing
  • Choose from a range of sizes with a variety of connection options
  • Easily connect fluid paths without the use of tools
  • Male and female luers available
Specifications

Specifications

ISO 9001
Polycarbonate
Transitions between luer fittings and UNF or NPT threads
10 to 32 UNF
Luer Fitting
Female luer x 10 to 32 in. UNF thread
25 Pack
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.