missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha Packaging™ Polyethylene Biohazard Bags
Use either plain or printed Alpha Packaging™ Polyethylene Biohazard Bags for specimen collection and transport. They are printed with a biohazard warning.
Brand: Alpha Packaging™ 12390529
Description
- Plain or printed with a biohazard warning
- Opening along width. 50μm thickness
- Resealable section for the specimen
Specifications
Biohazard Bag | |
Polyethylene | |
50 μm | |
150 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction