Learn More
Kartell™ Polyethylene Disc Membrane
These discs are made of polyethylene and have good chemical resistance. Kartell™ Polyethylene Disc Membrane is used to separate the filter paper from the funnel plate when working with a Buchner Funnel.
Brand:  Kartell™ 0083800
9.13 EUR valid until 2024-12-31
Use promo code "21931" to get your promotional price.
 View more versions of this product
Product Code. 10001051
  • 10.05 € / Pack of 10
Estimated Shipment: 02-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Useful interface between filter paper and perforated Buchner plate
  • Avoids clogging of filter holes
Specifications
Specifications
Disc Membrane
Polyethylene
300 μm
10/Pack
Disc Membrane
Polyethylene
Interface between filter paper and perforated Buchner plate
80 mm