Learn More
asecos™ Polyethylene Drum Chock
The Polyethylene Drum Chock is ideal for the safe emptying of drums.
Brand:  asecos™ 2802
 View more versions of this product
Product Code. 17816476
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • Seal made of EPDM
  • For caustic and aggressive chemicals
Specifications
Specifications
Brown
Polyethylene
162.00 mm
156.00 mm
0.5 kg
2 in. Drum
6.38 in.
6.14 in.
1.10 lb.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
asecos™ Polyethylene Drum Chock > 2 in. Threads