missing translation for 'onlineSavingsMsg'
Learn More

Oakton™ Glass Beads for Mills

Suitable glass beads for the mini-BeedBeater™ Mill. Cole-Parmer™ Glass Beads for Mills are used to crush and separate small samples from bone to microbial spores.

Brand:  Oakton™ 36270-51

 View more versions of this product

Product Code. 12614267

  • 96.45 € / Each
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Suitable glass beads for the mini-BeedBeater™ Mill. Cole-Parmer™ Glass Beads for Mills are used to crush and separate small samples from bone to microbial spores.

  • Available in three different sizes
  • Crush samples such as bone and microbial spores
  • Handles small samples efficiently—from 100μL to 1μL
  • Non-foaming, non-aerosol method preserves enzymes, nucleic acids, and organelles
Specifications

Specifications

Beads
0.5 mm
1 lb.
Glass
Mini-Bead Beater Glass Mill
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Oakton™ Glass Beads for Mills > Diameter: 0.5mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.