Learn More
Masterflex™ Polyethylene Male Pipe Adapter
Designed for quick and easy assembly without tools
Brand:  Masterflex™ 30621-26
 View more versions of this product
Product Code. 11736008
  • 20.50 € / Pack of 10
Estimated Shipment: 21-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polyethylene Male Pipe Adapter
  • Adapts from a NPT male thread to a hose barb
  • Barbed fittings are designed for quick and easy assembly without tools
  • Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30psi)
  • Available in packs of ten
Specifications
Specifications
Barb x NPT(M) Connection
HDPE
1/8 in.
1/8 in.
1/8 x 1/8 in.
Male Pipe Adapter
Not Autoclavable
10 Fittings
1/8 in.
Ethylene Oxide
Straight