missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Masterflex™ Polyethylene Male Pipe Adapter
Designed for quick and easy assembly without tools
20.50 € - 29.15 €
Specifications
Product Type | Male Pipe Adapter |
---|---|
Description | Barb x NPT(M) Connection |
Autoclavable | Not Autoclavable |
Material | HDPE |
Sterilization Method | Ethylene Oxide |
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
11736008
|
Masterflex™
30621-26 |
1/8 x 1/8 in. |
20.50 €
Pack of 10 |
Estimated Shipment: 04-03-2025
Log in to see stock. |
|||||
11735988
|
Masterflex™
30621-48 |
1/4 x 5/16 in. |
25.10 €
Pack of 10 |
Estimated Shipment: 04-03-2025
Log in to see stock. |
|||||
11765259
|
Masterflex™
30621-12 |
3/4 x 3/4 in. |
28.25 €
Pack of 10 |
Estimated Shipment: 13-03-2025
Log in to see stock. |
|||||
11705489
|
Masterflex™
30621-54 |
1/4 x 1/2 in. |
24.85 €
Pack of 10 |
Estimated Shipment: 13-03-2025
Log in to see stock. |
|||||
11780389
|
Masterflex™
30621-66 |
1/2 x 5/8 in. |
29.15 €
Pack of 10 |
Estimated Shipment: 13-03-2025
Log in to see stock. |
|||||
11795539
|
Masterflex™
30621-70 |
1/2 x 3/4 in. |
29.15 €
Pack of 10 |
Estimated Shipment: 13-03-2025
Log in to see stock. |
|||||
Description
- Polyethylene Male Pipe Adapter
- Adapts from a NPT male thread to a hose barb
- Barbed fittings are designed for quick and easy assembly without tools
- Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30psi)
- Available in packs of ten
Specifications
Male Pipe Adapter | |
Not Autoclavable | |
Ethylene Oxide |
Barb x NPT(M) Connection | |
HDPE |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title