missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Sterile Polyethylene Sampling Bags

Designed with a twirl round metal wire closure system and puncture proof safety tabs. Fisherbrand™ Sterile Polyethylene Sampling Bags are leakproof when sealed.

Brand:  Fisherbrand™ EPL-3050-FS-1_OLD

Product Code. 11512773

  • 161.20 € / Pack of 1000
Estimated Shipment: 25-06-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Manufactured with pliant, highly resistant virgin polyethylene 
  • Sterile transparent polyethylene bags without side seals
  • Choose clear or white marking area for written notations
  • Convenient pull tabs to open bag without touching the edges
  • Ideal for use with solid, semi-solid or liquid samples
  • For use in all laboratories where samples need to be collected into a sterile environment.
Specifications

Specifications

60 mL
Use in all laboratories where samples need to be collected into a sterile environment
Polyethylene
Sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Sterile Polyethylene Sampling Bags > Capacity: 60mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.