missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ Wide Neck Theraphthalate (PETG) Containers, Crystal-clear

Use the brilliant transparency of the Kautex™ Wide Neck Theraphthalate (PETG) Containers, Crystal-clear for a great product presentation.

Brand:  Kautex™ 2000074373

 View more versions of this product

Product Code. 17878127

  • 667.00 € / Pack of 406

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Series 310
  • Transparent PETG
  • Square with wide neck opening
  • Easy to fill and clean
  • Leakproof
  • Without closure
  • Ideal for product presentation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

200 mL
41 mm
Closure, Foam Insert
406
Black
Square
Wide Neck Container
Polyethylene
Transparent
Packaging, Dispensing, Sending Samples, Storage, Mailing
PET
406/Cs.
No
88 mm
Closure, Foam Insert
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.