missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GUERIN PLASTIQUES Polyethylene Sleeves
Use for food handling and general applications. GUERIN PLASTIQUES Polyethylene Sleeves are made of transparent, 0.1mm thick polyethylene.
Brand: GUERIN PLASTIQUES 50152
This item is not returnable.
View return policy
Description
- 350m long in total
- Available in a variety of widths
Specifications
1/Pk. | |
Polyethylene | |
350 m | |
Transparent |
Sleeve | |
Food Quality | |
0.1 Thick x 100Wmm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GUERIN PLASTIQUES Polyethylene Sleeves > Size: 0.1 Thick x 100Wmm
Spot an opportunity for improvement?Share a Content Correction