missing translation for 'onlineSavingsMsg'
Learn More

GUERIN PLASTIQUES Polyethylene Sleeves

Use for food handling and general applications. GUERIN PLASTIQUES Polyethylene Sleeves are made of transparent, 0.1mm thick polyethylene.

Brand:  GUERIN PLASTIQUES 50155

 View more versions of this product

Product Code. 11836523

  • 278.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 350m long in total
  • Available in a variety of widths
Specifications

Specifications

1/Pk.
Polyethylene
350 m
Transparent
Sleeve
Food Quality
0.1 Thick x 160Wmm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
GUERIN PLASTIQUES Polyethylene Sleeves > Size: 0.1 Thick x 160Wmm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.