Learn More
Polyethyleneimine, branched, M.W. 50,000 - 100,000, 30% w/w aq. soln., Thermo Scientific Chemicals
66.83 € - 798.98 €
Chemical Identifiers
CAS | 9002-98-6 |
---|---|
Molecular Formula | ((C2H5N)zC2H4N)y(C2H5N)x |
Molecular Weight (g/mol) | 43.07 |
MDL Number | MFCD00084427 |
InChI Key | NOWKCMXCCJGMRR-UHFFFAOYSA-N |
Synonym | ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin |
PubChem CID | 9033 |
ChEBI | CHEBI:30969 |
IUPAC Name | aziridine |
SMILES | *-CCNCCN(-*)CCN-* |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15436675
|
Thermo Scientific Alfa Aesar
045024.14 |
25 g |
66.83 €
25g |
In Stock
Log in to see stock. |
|||||
15446675
|
Thermo Scientific Alfa Aesar
045024.22 |
100 g |
192.96 €
100g |
In Stock
Log in to see stock. |
|||||
15456675
|
Thermo Scientific Alfa Aesar
045024.36 |
500 g |
798.98 €
500g |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||||
Description
Polyethyleneimine acts as a protein precipitant used to purify proteins. It is used as a chelating agent and as a scavenger for aldehydes and oxides. It is also used in detergents, paper industry, dyes, printing inks and in water treatment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
9002-98-6 | |
43.07 | |
NOWKCMXCCJGMRR-UHFFFAOYSA-N | |
9033 | |
aziridine |
((C2H5N)zC2H4N)y(C2H5N)x | |
MFCD00084427 | |
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin | |
CHEBI:30969 | |
*-CCNCCN(-*)CCN-* |
Specifications
Colorless to Yellow | |
Liquid | |
MFCD00084427 | |
Soluble in hot water at a low pH,methanol,ethanol,or chloroform. Insoluble in cold water,benzene,ethyl ether and acetone. | |
*-CCNCCN(-*)CCN-* | |
43.07 | |
CHEBI:30969 | |
Odorless |
9002-98-6 | |
((C2H5N)zC2H4N)y(C2H5N)x | |
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin | |
NOWKCMXCCJGMRR-UHFFFAOYSA-N | |
aziridine | |
9033 | |
30% w/w aq. soln. | |
Polyethyleneimine, Branched, M.W. 50,000 to 100,000 |
Safety and Handling
missing translation for 'rtecsNumber' : TQ7915000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.