Learn More
Macherey-Nagel™ Polygoprep™ 1000 to 30 C18
Brand:  Macherey-Nagel™ 711029.100
 View more versions of this product
Product Code. 10388341
  • 414.00 € / 100g
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
40 to 63 μm
POLYGOPREP 1000 to 30
0.1 kg
Polygoprep
1000 Å
2 to 8
POLYGOPREP C18
Liquid Chromatography