Learn More
Macherey-Nagel™ Polygoprep™ 1000 to 30 C18
414.00 € - 3245.00 €
Specifications
Pore Size 1000 Å
Stationary Phase POLYGOPREP 1000 to 30
pH 2 to 8
Sorbent POLYGOPREP C18
Type Polygoprep
View More Specs
Products 4
Product Code Brand Quantity Particle Size Price Quantity & Availability  
Product Code Brand Quantity Particle Size Price Quantity & Availability  
10787842
View Documents
Macherey-Nagel™
711028.100
0.1 kg 25 to 40 μm
428.00 €
100g
Estimated Shipment: 13-02-2025
to see stock.
Add to basket
10388341
View Documents
Macherey-Nagel™
711029.100
0.1 kg 40 to 63 μm
414.00 €
100g
Estimated Shipment: 13-02-2025
to see stock.
Add to basket
10526131
View Documents
Macherey-Nagel™
711028.1000
1 kg 25 to 40 μm
3245.00 €
1000g
Estimated Shipment: 13-02-2025
to see stock.
Add to basket
10788032
View Documents
Macherey-Nagel™
711029.1000
1 kg 40 to 63 μm
3145.00 €
1000g
Estimated Shipment: 13-02-2025
to see stock.
Add to basket
Specifications
1000 Å
2 to 8
Polygoprep
POLYGOPREP 1000 to 30
POLYGOPREP C18
Liquid Chromatography