Learn More
Macherey-Nagel™ Polygoprep™ 1000 to 30 C18
Brand:  Macherey-Nagel™ 711028.1000
 View more versions of this product
Product Code. 10526131
  • 3245.00 € / 1000g
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
25 to 40 μm
POLYGOPREP 1000 to 30
1 kg
Polygoprep
1000 Å
2 to 8
POLYGOPREP C18
Liquid Chromatography