Learn More
Polypropylene Disposal Bins
146.00 € - 287.00 €
Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
13433657
View Documents
Daniels Healthcare
DD672
2.5 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
13101264
View Documents
Daniels Healthcare
DD673
7 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Ideal for cytotoxic and cytostatic waste which, to meet regulations, should be disposed of separately from other healthcare waste. The SHARPSGUARD™ cyto lid assists safe, easy collection of items including giving sets, syringes and gloves. Color-coded with a purple lid, handle and label, it can be safely transported for incineration after closure. Absorbent pads are included. The container is made to BS EN ISO 23907.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Polypropylene Disposal Bins