missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Polypropylene Flask Holder

These unique holders are perfect for keeping round and tapered bottom flasks upright.

Brand:  Bel-Art™ SP Scienceware™ F38951-2010

 View more versions of this product

Product Code. 10406162

  • 45.65 € / Pack of 3
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Inside wall has a slight taper to the open bottom; allows a variety of flask shapes and sizes to fit into one holder
  • Wide base keeps flasks stable
  • Side slots can be used as sight windows
  • Molded from polypropylene plastic; durable and easy to clean
  • Autoclavable at 121°C (250°F)
Specifications

Specifications

Flask Holder
Red
Polypropylene
121°C
Autoclavable
Round Bottom Flask (250 mL)
3 Pack
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.