missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Scienceware™ Polypropylene Funnel Holder

Non-corrosive polypropylene plastic funnel holder and clamp supports several funnel sizes.

Brand:  Bel-Art™ Scienceware™ F18242-0000

Product Code. 10595994

This item is not returnable. View return policy

Description

Description

  • Outer ring supports 76 to 152mm top diameter funnels
  • Removable inner ring holds smaller funnels from 25 to 76mm top diameter
  • Includes two bushings for 12.7mm or 9.5mm diameter support rods
Specifications

Specifications

Two bushings for 12.7mm (1/2 in.) or 9.5mm (3/8 in.) diameter support rods.
Double
Holds 25mm to 152mm funnels
Funnel Holder
Polypropylene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ Scienceware™ Polypropylene Funnel Holder > For use with: Holds 25mm to 152mm funnels

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.