missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ Polypropylene Funnel Holders
Manufactured from polypropylene for durability and strength. Kartell™ Polypropylene Funnel Holders support a variety of funnel types and sizes.
Brand: Kartell™ 0097000
8.52 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
Manufactured from polypropylene for durability and strength. Kartell™ Polypropylene Funnel Holders support a variety of funnel types and sizes.
- Supplied with two olives for tight fit on supporting rods
- Available in horseshoe style for larger separating funnels and circular style for small funnels
- Circular holder can be used with powder funnels with stem diameter not exceeding 35mm
- Horseshoe holder
Specifications
Horseshoe funnel holder | |
Polypropylene | |
One |
supporting rod and funnels, particularly separating funnels, from 125 to 1000mm diameter | |
2 olives for use with rods from 8 to 14mm | |
1 Each |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction