Learn More
Brady™ Polypropylene Label Tape for BradyJet J4000
This B-7425J polypropylene material is ideal for GHS and other full-color identification applications where resistance to harsh chemicals is required.
Brand:  Brady™ 152785
 View more versions of this product
Product Code. 17469222
  • 595.00 € / Each
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Matte white polypropylene with acrylic adhesive is Inkjet printable
  • Low-cost, full-color GHS labeling option withstands chemical exposure
  • Service temperature of -80°C to 70°C
  • An ink cartridge is required to print this material
Specifications
Specifications
Autoclavable
Permanent Acrylic
Matte
Ink Jet
Hazard Communication Identification, Equipment Labeling, Laboratory Labeling, General Identification
1
-80°C to +70°C
30.5 m
J40C-6000-7425J
White
Polypropylene (Inkjet-receptive)
Non-sterile
BradyJet J4000 Color Label Printer
1 Tape
0.13 mm
152.4 mm