missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Polypropylene Label Tape for BradyJet J4000
This B-7425J polypropylene material is ideal for GHS and other full-color identification applications where resistance to harsh chemicals is required.
296.00 € - 723.00 €
Specifications
Autoclavable | Autoclavable |
---|---|
Adhesive Type | Permanent Acrylic |
Color | White |
Finish | Matte |
Material | Polypropylene (Inkjet-receptive) |
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | |||||
17449222
|
Brady™
152783 |
57.1 mm |
296.00 €
Each |
Estimated Shipment: 25-02-2025
Log in to see stock. |
|||||
17459222
|
Brady™
152784 |
101.6 mm |
367.00 €
Each |
Estimated Shipment: 25-02-2025
Log in to see stock. |
|||||
17469222
|
Brady™
152785 |
152.4 mm |
595.00 €
Each |
Estimated Shipment: 25-02-2025
Log in to see stock. |
|||||
17479222
|
Brady™
152786 |
203.2 mm |
723.00 €
Each |
Estimated Shipment: 25-02-2025
Log in to see stock. |
|||||
Description
- Matte white polypropylene with acrylic adhesive is Inkjet printable
- Low-cost, full-color GHS labeling option withstands chemical exposure
- Service temperature of -80°C to 70°C
- An ink cartridge is required to print this material
Specifications
Autoclavable | |
White | |
Polypropylene (Inkjet-receptive) | |
Non-sterile | |
BradyJet J4000 Color Label Printer | |
1 Tape | |
0.13 mm |
Permanent Acrylic | |
Matte | |
Ink Jet | |
Hazard Communication Identification, Equipment Labeling, Laboratory Labeling, General Identification | |
1 | |
-80°C to +70°C | |
30.5 m |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title