Learn More
Masterflex™ Polypropylene Luer Fitting
Featuring a variety of fitting options, these Luer fittings are ideal for many applications including syringes or small tubing. Cole-Parmer™ Polypropylene Luer Fitting are available with standard, wide, and low-profile barbs.
Brand: Masterflex™ 45508-66
This item is not returnable.
View return policy
Description
- Three barb styles: standard, wide-bore, and low-profile.
- Wide-bore barbs offer enhanced fluid flow through the fitting and are usable with any flexible tubing.
- Low-profile barbs are ideal for use with semi-rigid tubing, such as nylon, polyethylene, or polypropylene.
- Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable.
- All parts meet tight dimensional tolerances for increased reliability and process purity.
- Choose from a range of sizes, 1/16in. to 1/4in., with a variety of connection options.
- Pack of 25
Specifications
ISO 594-1 standards | |
Polypropylene | |
25 Pack |
Luer Fitting | |
Female luer x 1/4 to 28 in. UNF thread |